5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2,3-dihydro-7-azaindole
073184-1G
073184-5G
073184-10G
102528-828EA
427.69
USD
102528-828
102528-830
102528-832
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₇H₇BrN₂ MW: 199.0479 g/mol |
MDL Number:
MFCD06659750 CAS Number: 115170-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...