5-Bromo-1H-pyrazolo[3,4-c]pyridine ≥95%

Supplier: Matrix Scientific

073183-1G 073183-5G
102528-824EA 638.46 USD
102528-824 102528-826
5-Bromo-1H-pyrazolo[3,4-c]pyridine ≥95%
5-Bromo-1H-pyrazolo[3,4-c]pyridine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD11518975
CAS Number: 929617-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR