6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

073180-250MG 073180-1G 073180-5G
102528-806EA 2692.31 USD
102528-806 102528-808 102528-810
6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥95%
6-Bromo-8-chloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄BrClN₂
MDL Number: MFCD11518983
CAS Number: 474708-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR