6-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

073179-250MG 073179-1G 073179-5G
102528-800EA 415.77 USD
102528-800 102528-802 102528-804
6-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥95%
6-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD11518940
CAS Number: 220896-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR