4,6-Dichloroimidazo[4,5-c]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
4,6-Dichloro-1H-imidazo[4,5-c]pyridine
073176-250MG
073176-1G
073176-5G
102528-784EA
740
USD
102528-784
102528-786
102528-788
4,6-Dichloroimidazo[4,5-c]pyridine ≥95%
4,6-Dichloroimidazo[4,5-c]pyridine
Hazard: Irritant
Formula:
C₆H₃Cl₂N₃ MW: 188.0141 g/mol |
MDL Number:
MFCD09753581 CAS Number: 2589-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...