3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride ≥95%

Supplier: Matrix Scientific

073175-1G 073175-5G
102528-778EA 32.31 USD
102528-778 102528-780
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride ≥95%
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride

Hazard: Irritant

Formula: C6H8ClF3N4
Melting Pt: 264 °C (dec.)
MDL Number: MFCD09817638
CAS Number: 762240-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR