2,4-Dichloropyrido[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073174-250MG 073174-1G 073174-5G
102528-772EA 450.42 USD
102528-772 102528-774 102528-776
2,4-Dichloropyrido[3,2-d]pyrimidine ≥95%
2,4-Dichloropyrido[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₃Cl₂N₃
MW: 200.0248 g/mol
MDL Number: MFCD11046863
CAS Number: 39551-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR