(1-Tosyl-1H-pyrrolo[2,3-b]pyridin-3-yl)boronic acid ≥95%

Supplier: Matrix Scientific

073173-250MG 073173-1G 073173-5G
102528-766EA 924.62 USD
102528-766 102528-768 102528-770
(1-Tosyl-1H-pyrrolo[2,3-b]pyridin-3-yl)boronic acid ≥95%
(1-Tosyl-1H-pyrrolo[2,3-b]pyridin-3-yl)boronic acid

Hazard: Irritant

Formula: C₁₄H₁₃BN₂O₄S
MW: 316.14 g/mol
MDL Number: MFCD10696634
CAS Number: 882562-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR