1,3-Difluoro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoronitrobenzene

073172-100G 073172-25G
102528-764EA 507.69 USD
102528-764 102663-936
1,3-Difluoro-2-nitrobenzene ≥95%
1,3-Difluoro-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 91…92 °C (11 mmHg)
Density: 1.503
MDL Number: MFCD00192035
CAS Number: 19064-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR