2,6-Dichloro-4-hydroxypyridine ≥95%

Supplier: Matrix Scientific

073171-1G 073171-5G 073171-10G
102528-754EA 710.77 USD
102528-754 102528-756 102528-758
2,6-Dichloro-4-hydroxypyridine ≥95%
2,6-Dichloro-4-hydroxypyridine

Hazard: Irritant

Formula: C₅H₃Cl₂NO
MW: 163.9894 g/mol
MDL Number: MFCD00955619
CAS Number: 17228-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR