5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

073170-1G 073170-5G 073170-10G
102528-748EA 213.85 USD
102528-748 102528-750 102528-752
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥95%
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.2118 g/mol
MDL Number: MFCD01464030
CAS Number: 4242-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR