tert-Butyl-4-carbamothioylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Carbamothioylpiperidine, N1-BOC protected, tert-Butyl 4-carbamothioylpiperidine-1-carboxylate

073168-5G 073168-25G
102528-738EA 218.46 USD
102528-738 102833-608
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂S
MW: 244.3500 g/mol
MDL Number: MFCD02180954
CAS Number: 214834-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR