5,6,7,8-Tetrahydro-1,6-naphthyridine ≥95%

Supplier: Matrix Scientific

073167-250MG 073167-1G
102528-732EA 946.15 USD
102528-732 102528-734
5,6,7,8-Tetrahydro-1,6-naphthyridine ≥95%
5,6,7,8-Tetrahydro-1,6-naphthyridine

Hazard: Irritant

Formula: C₈H₁₀N₂
MW: 134.1800 g/mol
MDL Number: MFCD06738733
CAS Number: 80957-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR