3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine ≥95%

Supplier: Matrix Scientific

073166-250MG 073166-1G
102528-728EA 853.85 USD
102528-728 102528-730
3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine ≥95%
3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine

Hazard: Irritant

Formula: C9H13N3
MW: 163.2196 g/mol
MDL Number: MFCD09743968
CAS Number: 216966-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR