tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 7-amino-3,4-dihydroquinoline-1(2H)-carboxylate

073165-250MG 073165-1G 073165-5G
102528-722EA 498.46 USD
102528-722 102528-724 102528-726
tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate ≥95%
tert-Butyl-7-amino-3,4-dihydroquinoline-1(2H)-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MW: 248.3200 g/mol
MDL Number: MFCD10566742
CAS Number: 351324-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR