1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

073162-250MG 073162-1G 073162-5G
102528-706EA 450.42 USD
102528-706 102528-708 102528-710
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid ≥95%
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD11043014
CAS Number: 53484-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR