Ethyl-4-aminopyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-aminopyrimidine-2-carboxylate

073160-1G 073160-5G
102528-696EA 687.69 USD
102528-696 102528-698
Ethyl-4-aminopyrimidine-2-carboxylate ≥95%
Ethyl-4-aminopyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
MDL Number: MFCD11973676
CAS Number: 71470-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR