3-Amino-4-methoxypyridine ≥95%

Supplier: Matrix Scientific

073156-1G 073156-5G
102528-674EA 163.08 USD
102528-674 102528-676
3-Amino-4-methoxypyridine ≥95%
3-Amino-4-methoxypyridine

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD07374959
CAS Number: 33631-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR