Azetidine-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: azetidine-2-carboxylic acid

073153-1G 073153-5G
102528-660EA 69.23 USD
102528-660 102528-662
Azetidine-2-carboxylic acid ≥98%
Azetidine-2-carboxylic acid

Hazard: Irritant

Formula: C₄H₇NO₂
Melting Pt: 217 °C (dec.)
MDL Number: MFCD00066660
CAS Number: 2517-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR