2,4,6-Trichloro-5-cyanopyrimidine ≥95%

Supplier: Matrix Scientific

073151-1G 073151-5G
102528-652EA 604.62 USD
102528-652 102528-654
2,4,6-Trichloro-5-cyanopyrimidine ≥95%
2,4,6-Trichloro-5-cyanopyrimidine

Hazard: Irritant

Formula: C₅Cl₃N₃
MDL Number: MFCD07636745
CAS Number: 3029-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR