5-Fluoropyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-fluoropyrimidine

073150-250MG 073150-1G 073150-5G
102528-646EA 250.11 USD
102528-646 102528-648 102528-650
5-Fluoropyrimidin-2-amine ≥95%
5-Fluoropyrimidin-2-amine

Hazard: Irritant

Formula: C4H4FN3
Melting Pt: 192…193 °C
MDL Number: MFCD11846500
CAS Number: 1683-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR