(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-butyl 3-methyl (3S)-piperazine-1,3-dicarboxylate

073149-1G 073149-5G
102528-642EA 166.15 USD
102528-642 102528-644
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate ≥95%
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₄
MW: 244.2875 g/mol
MDL Number: MFCD04115326
CAS Number: 314741-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR