3-Aminopyridine-4-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-cyanopyridine

073148-250MG 073148-1G 073148-5G
102528-636EA 85.54 USD
102528-636 102528-638 102528-640
3-Aminopyridine-4-carbonitrile ≥95%
3-Aminopyridine-4-carbonitrile

Hazard: Toxic

Formula: C₆H₅N₃
MW: 119.13 g/mol
Melting Pt: 135…138 °C
MDL Number: MFCD04035596
CAS Number: 78790-79-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR