7-Methyl-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

073145-250MG 073145-1G 073145-5G
102528-620EA 427.69 USD
102528-620 102528-622 102528-624
7-Methyl-1,2,3,4-tetrahydroisoquinoline ≥95%
7-Methyl-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₀H₁₃N
MW: 147.2200 g/mol
MDL Number: MFCD03004622
CAS Number: 207451-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR