5-Fluoro-2-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

073143-5G 073143-10G 073143-100G
102528-608EA 143.08 USD
102528-608 102528-610 102528-612
5-Fluoro-2-methylbenzaldehyde ≥95%
5-Fluoro-2-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₇FO
MDL Number: MFCD02261763
CAS Number: 22062-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR