2-Chloro-4-methyl-5-pyridineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Chloro-4-methylpyridin-3-yl)boronic acid, 2-Chloro-4-methylpyridine-5-boronic acid

073142-1G 073142-5G 073142-10G
102528-602EA 198.46 USD
102528-602 102528-604 102528-606
2-Chloro-4-methyl-5-pyridineboronic acid ≥95%
2-Chloro-4-methyl-5-pyridineboronic acid

Hazard: Irritant

Formula: C₆H₇BClNO₂
MW: 171.3900 g/mol
MDL Number: MFCD07368882
CAS Number: 913836-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR