4-Amino-5-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073140-250MG 073140-1G 073140-5G
102528-592EA 640 USD
102528-592 102528-594 102528-596
4-Amino-5-methylpyrimidine ≥95%
4-Amino-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.1300 g/mol
MDL Number: MFCD09909807
CAS Number: 22433-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR