6-Bromobenzo[d]oxazole ≥95%
Supplier: Matrix Scientific
073139-1G
073139-5G
102528-588EA
330.77
USD
102528-588
102528-590
6-Bromobenzo[d]oxazole ≥95%
6-Bromobenzo[d]oxazole
Hazard: Irritant
Formula:
C₇H₄BrNO MW: 198.0168 g/mol |
MDL Number:
MFCD03092891 CAS Number: 375369-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...