1-Amino-7-bromoisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Bromoisoquinolin-1-amine

073136-250MG 073136-1G
102528-570EA 684.62 USD
102528-570 102528-572
1-Amino-7-bromoisoquinoline ≥95%
1-Amino-7-bromoisoquinoline

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.0700 g/mol
MDL Number: MFCD07374397
CAS Number: 215453-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR