2-Bromoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

073135-1G 073135-5G
102528-566EA 372.31 USD
102528-566 102528-568
2-Bromoimidazo[1,2-a]pyridine ≥95%
2-Bromoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD09834778
CAS Number: 112581-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR