7-Bromo-1H-pyrrolo[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

073133-250MG 073133-1G
102528-554EA 347.69 USD
102528-554 102528-556
7-Bromo-1H-pyrrolo[3,2-c]pyridine ≥95%
7-Bromo-1H-pyrrolo[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD07781153
CAS Number: 902837-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR