5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

073132-5G 073132-25G
102528-550EA 40 USD
102528-550 102833-606
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid ≥95%
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₅ClN₂O₂S
MW: 204.6300 g/mol
MDL Number: MFCD00173907
CAS Number: 61727-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR