7-Hydroxy-2-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Hydroxy-3,4-dihydro-1H-naphthalen-2-one

073131-1G 073131-5G
102528-542EA 201.54 USD
102528-542 102528-544
7-Hydroxy-2-tetralone ≥95%
7-Hydroxy-2-tetralone

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MDL Number: MFCD04038657
CAS Number: 37827-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR