6-Bromobenzo[b]thiophene ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromobenzothiophene

073130-250MG 073130-1G 073130-5G
102528-536EA 355.38 USD
102528-536 102528-538 102528-540
6-Bromobenzo[b]thiophene ≥95%
6-Bromobenzo[b]thiophene

Hazard: Irritant

Formula: C₈H₅BrS
MW: 213.0943 g/mol
MDL Number: MFCD11226847
CAS Number: 17347-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR