3-Bromoimidazo[1,2-b]pyridazine ≥95%

Supplier: Matrix Scientific

073128-1G 073128-5G
102528-526EA 36.92 USD
102528-526 102528-528
3-Bromoimidazo[1,2-b]pyridazine ≥95%
3-Bromoimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C6H4BrN3
MW: 198.0201 g/mol
MDL Number: MFCD09757672
CAS Number: 18087-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR