Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate

073127-100G 073127-25G
102528-524EA 76.92 USD
102528-524 102663-932
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥95%
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₉NO₄
MW: 253.30 g/mol
Melting Pt: 178…183 °C
MDL Number: MFCD00005951
CAS Number: 1149-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR