N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-4,6-dichloro-5-formamidopyrimidine, N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide
073126-5G
073126-10G
073126-100G
102528-516EA
38.46
USD
102528-516
102528-518
102528-520
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥95%
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide
Hazard: Irritant
Formula:
C₅H₄Cl₂N₄O MW: 207.0200 g/mol |
MDL Number:
MFCD04112936 CAS Number: 171887-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...