3,5-Dibromoisonicotinaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Dibromo-4-formylpyridine, 3,5-Dibromo-4-pyridinecarboxaldehyde, 3,5-Dibromoisonicotinaldehyde
073119-5G
073119-25G
102528-478EA
67.69
USD
102528-478
102833-604
3,5-Dibromoisonicotinaldehyde ≥95%
3,5-Dibromoisonicotinaldehyde
Hazard: Toxic/Stored under Argon/Light Sensitive/Air Sensitive
Formula:
C6H3Br2NO Melting Pt: 118…122 °C |
MDL Number:
MFCD06410681 CAS Number: 70201-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...