2-Methyl-m-anisic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methoxy-2-methylbenzoic acid
073117-100G
073117-25G
102528-468EA
189.23
USD
102528-468
102833-600
2-Methyl-m-anisic acid ≥95%
2-Methyl-m-anisic acid
Hazard: Irritant
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Number:
MFCD02094039 CAS Number: 55289-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...