4-Bromo-3-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
073112-5G
073112-25G
102528-440EA
83.08
USD
102528-440
102833-598
4-Bromo-3-nitrobenzaldehyde ≥95%
4-Bromo-3-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₇H₄BrNO₃ |
MDL Number:
MFCD00204031 CAS Number: 163596-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...