6-Cyano-2-naphthol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Cyano-2-naphthol, 6-Hydroxy-2-naphthonitrile

073106-1G 073106-5G 073106-10G
102528-404EA 15.38 USD
102528-404 102528-406 102528-408
6-Cyano-2-naphthol ≥95%
6-Cyano-2-naphthol

Hazard: Irritant

Formula: C₁₁H₇NO
MW: 169.1794 g/mol
MDL Number: MFCD01863480
CAS Number: 52927-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR