1-Chloro-2,3-difluoro-5-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4,5-difluoronitrobenzene

073105-1G 073105-5G
102528-400EA 364.62 USD
102528-400 102528-402
1-Chloro-2,3-difluoro-5-nitrobenzene ≥95%
1-Chloro-2,3-difluoro-5-nitrobenzene

Hazard: Irritant

Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
MDL Number: MFCD06658261
CAS Number: 53780-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR