Benzo[b]thiophene-3-carboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: Benzo[b]thiophene-3-carbaldehyde

073096-1G 073096-5G
102528-354EA 38.46 USD
102528-354 102528-356
Benzo[b]thiophene-3-carboxaldehyde ≥95%
Benzo[b]thiophene-3-carboxaldehyde

Hazard: Irritant

Formula: C₉H₆OS
MW: 162.2083 g/mol
MDL Number: MFCD00052376
CAS Number: 5381-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR