5-Methylpyrazine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
073092-100G
073092-25G
102528-336EA
153.85
USD
102528-336
102833-592
5-Methylpyrazine-2-carboxylic acid ≥95%
5-Methylpyrazine-2-carboxylic acid
Hazard: Irritant
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00068241 CAS Number: 5521-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...