(1H,3H)-Quinazoline-2,4-dione ≥95%
Supplier: Matrix Scientific
Synonyms:
Quinazoline-2,4(1H,3H)-dione, Benzoyleneurea
073089-5G
073089-10G
073089-100G
102528-320EA
64.62
USD
102528-320
102528-322
102528-324
(1H,3H)-Quinazoline-2,4-dione ≥95%
(1H,3H)-Quinazoline-2,4-dione
Hazard: Irritant
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol |
MDL Number:
MFCD00006699 CAS Number: 86-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...