4-Nitro-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-2-methylaniline, 2-Methyl-4-nitroaniline, 2-Amino-5-nitrotoluene

073088-10G 073088-100G
102528-316EA 36.92 USD
102528-316 102528-318
4-Nitro-o-toluidine ≥95%
4-Nitro-o-toluidine

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD00007734
CAS Number: 99-52-5
UN: 2660
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR