6-Bromo-3,4-dihydro-1H-quinolin-2-one ≥95%
Supplier: Matrix Scientific
073082-1G
073082-5G
073082-10G
102528-296EA
21.54
USD
102528-296
102528-298
102528-300
6-Bromo-3,4-dihydro-1H-quinolin-2-one ≥95%
6-Bromo-3,4-dihydro-1H-quinolin-2-one
Hazard: Irritant
Formula:
C₉H₈BrNO MW: 226.07 g/mol Melting Pt: 170…172 °C |
MDL Number:
MFCD03839832 CAS Number: 3279-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...