2-Methyl-5-pyridineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Methylpyridin-3-yl)boronic acid

073081-1G 073081-5G
102528-292EA 92.31 USD
102528-292 102528-294
2-Methyl-5-pyridineboronic acid ≥95%
2-Methyl-5-pyridineboronic acid

Hazard: Irritant

Formula: C₆H₈BNO₂
MW: 136.94 g/mol
Melting Pt: 90-100°C
MDL Number: MFCD06801736
CAS Number: 659742-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR