4-Methyl-1-phenylpyrazolidin-3-one 95

Supplier: Matrix Scientific

Synonyms: 4-Methyl-3-oxo-1-phenylpyrazolidine, 4-Methyl-1-phenyl-3-pyrazolidone

073078-5G 073078-10G
102528-276EA 90.77 USD
102528-276 102528-278
4-Methyl-1-phenylpyrazolidin-3-one 95
4-Methyl-1-phenylpyrazolidin-3-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.2151 g/mol
MDL Number: MFCD00044800
CAS Number: 2654-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR