3-Hydroxy-p-anisaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

073075-100G 073075-500G
102528-266EA 60 USD
102528-266 102663-926
3-Hydroxy-p-anisaldehyde ≥95%
3-Hydroxy-p-anisaldehyde

Hazard: Irritant/Light Sensitive/Air Sensitive

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 112…116 °C
MDL Number: MFCD00003369
CAS Number: 621-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR